Learn More
Alkali Scientific ASI™ 384 Black Well, Clear Bottom, Cell Culture Plate, Sterile, TC Treated, PS / FLux
SDP
Catalog No. 503027550
Encompass_Preferred
ASI™ 384 Well Cell Culture Plates provide high quality 384 HT Plates for high throughput research laboratories. Fluorescence and luminescence assays have been sucessfully tested with these 384 HT Plates, results featuring high signal with low background. Alphanuneric labeling provides easier well identification.  toal well volume = 120ul with a working well volume of 100ul. This STERILE polystyrene surface is positive charged & Cell Culture Treated - suitable for adherent cells Minimum light scattering and cross-talk during fluorescence assays HTS compatible Flat bottom Suitable for colorimetry, fluorescence and luminescence assay Transparent lid provided with each well plate 1 plate & lid per inner pack, total 10 plates/lids per CASE  
Catalog No. 50-302-7550 Supplier Alkali Scientific Supplier No. TPB3085
Add to Cart
Add to Cart
ASI™ 384 Well Cell Culture Plates provide high quality 384 HT Plates for high throughput research laboratories. Fluorescence and luminescence assays have been sucessfully tested with these 384 HT Plates, results featuring high signal with low background. Alphanuneric labeling provides easier well identification.  toal well volume = 120ul with a working well volume of 100ul. This STERILE polystyrene surface is positive charged & Cell Culture Treated - suitable for adherent cells Minimum light scattering and cross-talk during fluorescence assays HTS compatible Flat bottom Suitable for colorimetry, fluorescence and luminescence assay Transparent lid provided with each well plate 1 plate & lid per inner pack, total 10 plates/lids per CASE  