Learn More
SPT LABTECH INC 384 CH POLY REA RES,SW,185ML
Catalog No. NC1412485
Encompass
384 channel polypropylene reagent reservoir, Single well (185mL max volume)10 reservoirs/unit, 50 reservoirs/case
Catalog No. NC1412485 Supplier SPT LABTECH INC Supplier No. RES185384SW
384 channel polypropylene reagent reservoir, Single well (185mL max volume)10 reservoirs/unit, 50 reservoirs/case