Learn More
Alkali Scientific ASI™ 384 HT Cell Culture Plate, PS, 120μl,40/Cs, Clear, Unibinding
SDP
Catalog No. 503027595
Encompass_Preferred
ASI™ provides high quality 384 HT Plates for high throughput research laboratories. Fluorescence and luminescence assays have been successfully tested with SPL 384 HT Plates, results featuring high signal with low background.  ▫ White plates : maximum reflection and minimum cross-talk for luminescence assay.▫ Black plates  : minimum light scattering and cross-talk during fluorescence assay.▫ Maxibinding: Modified polystyrene surface for a higher binding capacity forproteins and other                      molecules with both hydrophilic and hydrophobic regions, such as immunoglobulins.                     (Recommended for most ELISA experiments)▫ Unibinding : This polystyrene surface is suitable for adsorption of hydrophobic molecules.                       (Antigen ELISA) ▪ 2 surface type▪ Suitable for colorimetry, fluorescence and luminescence assay▪ HTS (High-Throughput Screening) compatible▪ Transparent lid provided with each well plate▪ Flat bottom▪ Alphanumeric labeling
Catalog No. 50-302-7595 Supplier Alkali Scientific Supplier No. TPCU3084
ASI™ provides high quality 384 HT Plates for high throughput research laboratories. Fluorescence and luminescence assays have been successfully tested with SPL 384 HT Plates, results featuring high signal with low background.  ▫ White plates : maximum reflection and minimum cross-talk for luminescence assay.▫ Black plates  : minimum light scattering and cross-talk during fluorescence assay.▫ Maxibinding: Modified polystyrene surface for a higher binding capacity forproteins and other                      molecules with both hydrophilic and hydrophobic regions, such as immunoglobulins.                     (Recommended for most ELISA experiments)▫ Unibinding : This polystyrene surface is suitable for adsorption of hydrophobic molecules.                       (Antigen ELISA) ▪ 2 surface type▪ Suitable for colorimetry, fluorescence and luminescence assay▪ HTS (High-Throughput Screening) compatible▪ Transparent lid provided with each well plate▪ Flat bottom▪ Alphanumeric labeling