Learn More
Beckman Coulter Beckman Coulter Echo Qualified 384-Well Low Dead Volume (LDV) COC Source Microplate, Clear, Sterile - 50/Case
Catalog No. NC2980061
Encompass
Beckman Coulter Echo Qualified 384-well low dead volume (LDV) source microplates are engineered for acoustic liquid handling to minimize dead volume and enable precise, contactless transfer. Constructed from cyclic olefin copolymer (COC) with clear, flat-bottom wells and sterile packaging, these plates are optimized for acoustic performance, DMSO storage, heat sealing, and robotic workflows.
  • 384 wells, low dead volume design.
  • Constructed of cyclic olefin copolymer (COC) for optical clarity.
  • Clear, flat-bottom wells suitable for acoustic transfer.
  • Sterile packaging, 50 plates per case.
  • Compatible with heat sealing devices.
  • Suitable for DMSO storage.
  • Each lot tested and certified for performance.
Catalog No. NC2980061 Supplier Beckman Coulter Supplier No. 00116128
Beckman Coulter Echo Qualified 384-well low dead volume (LDV) source microplates are engineered for acoustic liquid handling to minimize dead volume and enable precise, contactless transfer. Constructed from cyclic olefin copolymer (COC) with clear, flat-bottom wells and sterile packaging, these plates are optimized for acoustic performance, DMSO storage, heat sealing, and robotic workflows.
  • 384 wells, low dead volume design.
  • Constructed of cyclic olefin copolymer (COC) for optical clarity.
  • Clear, flat-bottom wells suitable for acoustic transfer.
  • Sterile packaging, 50 plates per case.
  • Compatible with heat sealing devices.
  • Suitable for DMSO storage.
  • Each lot tested and certified for performance.