Learn More
Labsource Inc 4titude FrameStar 384-Well PCR Plate, Skirted, Polypropylene Wells, 50 Plates/Case
SDP
Catalog No. NC2383287
Encompass
The 4titude FrameStar 384-Well Skirted PCR Plate provides optimal performance in PCR and qPCR applications. Its dual-component design features thin-walled polypropylene wells for efficient thermal transfer and a rigid polycarbonate frame for stability and reduced evaporation. Tested in a cleanroom, it's certified free from contaminants like DNase and RNase, ensuring reliable results. This plate is compatible with standard thermal cyclers and automation systems, making it an ideal choice for labs seeking high-quality PCR supplies.
Catalog No. NC2383287 Supplier Labsource Inc Supplier No. P97115
Add to Cart
Add to Cart
The 4titude FrameStar 384-Well Skirted PCR Plate provides optimal performance in PCR and qPCR applications. Its dual-component design features thin-walled polypropylene wells for efficient thermal transfer and a rigid polycarbonate frame for stability and reduced evaporation. Tested in a cleanroom, it's certified free from contaminants like DNase and RNase, ensuring reliable results. This plate is compatible with standard thermal cyclers and automation systems, making it an ideal choice for labs seeking high-quality PCR supplies.