Learn More
Scinomix Inc 384 PIN REPLICATOR 120/CS
SDP
Catalog No. NC1286581
Encompass
The 384 Pin Replicator is a disposable replicator for one time use, that virtually eliminates the threat of cross-contamination from repeat use metal replicators. Our natural, unpigmented 384 Pin Replicator is molded from medical grade polypropylene . The parts were designed for robotics use and the top has approximately 5 mm of height on the portrait and landscape sides for easy gripping. Features of 384 Pin Replicators Pin Replicator Pin height: 12 mm 120 each per case, sold by the case
Catalog No. NC1286581 Supplier Scinomix Inc Supplier No. SCI6010OS
The 384 Pin Replicator is a disposable replicator for one time use, that virtually eliminates the threat of cross-contamination from repeat use metal replicators. Our natural, unpigmented 384 Pin Replicator is molded from medical grade polypropylene . The parts were designed for robotics use and the top has approximately 5 mm of height on the portrait and landscape sides for easy gripping. Features of 384 Pin Replicators Pin Replicator Pin height: 12 mm 120 each per case, sold by the case