Learn More
Analytical Sales & Services 384-Position Deep Well Reservoir with Two Controls Wells
Catalog No. 5000804789
Encompass
384-Position Deep Well Reservoir\n• 384-Position Base with Pyramid Bottoms\n• Two Control Wells\n• 282mL Total Plate Volume\n• Polypropylene\n\nQty/Pkg: 25
Catalog No. 50-008-04789 Supplier Analytical Sales & Services Supplier No. 38477
384-Position Deep Well Reservoir\n• 384-Position Base with Pyramid Bottoms\n• Two Control Wells\n• 282mL Total Plate Volume\n• Polypropylene\n\nQty/Pkg: 25