Learn More
Arcticwhite LLC Glass Coated 384 Square Well, 120 UL, Polypropylene, Flat Bottom, 384 Wells, Glass Coating (low-binding, Chemically Inert), 10 Microplates per Case
SDP
Catalog No. NC4032591
Encompass
Glass Coated 384 Square Well, 120 UL/Well, Polypropylene, Flat Bottom, 384 Wells, Glass Coating (low-binding, Chemically Inert), 10 Microplates per Case. A 384-well microplate for sample storage, compound management, and automated liquid handling. Format - 384 wells. Supplied by ArcticWhite as 10 microplates per case.
Catalog No. NC4032591 Supplier Arcticwhite LLC Supplier No. AWLSX11207
Glass Coated 384 Square Well, 120 UL/Well, Polypropylene, Flat Bottom, 384 Wells, Glass Coating (low-binding, Chemically Inert), 10 Microplates per Case. A 384-well microplate for sample storage, compound management, and automated liquid handling. Format - 384 wells. Supplied by ArcticWhite as 10 microplates per case.