Learn More
Analytical Sales & Services 384-Well 10 mil Adhesive Silicone Sealing Mat
Catalog No. 5000806406
Encompass
Advantage 384 Well, 10 mil Adhesive Silicone Sealing Mat; Gas Permeable, Breathable (Ideal for cell growth)\n\nQty/Pkg: 5
Catalog No. 50-008-06406 Supplier Analytical Sales & Services Supplier No. 38404
Advantage 384 Well, 10 mil Adhesive Silicone Sealing Mat; Gas Permeable, Breathable (Ideal for cell growth)\n\nQty/Pkg: 5