Learn More
Corning™ 384-Well Black/Clear Round Bottom Ultra-Low Attachment Spheroid Microplate with Generic Bar codes, With Lid, Sterile
GSA_VAGREENER_CHOICE
Catalog No. 13700560 Shop All Corning Products
Change view
Click to view available options
Color:
Black, Clear Bottom
Catalog No. Color
13-700-560 Black, Clear Bottom
1 options
Catalog No. 13-700-560 Supplier Corning™ Supplier No. 3830BC
Get Up to $2,500 in Products with Purchase
View all promotions
Add to Cart
Add to Cart
Ultra-Low Attachment microplates feature a covalently bound hydrogel layer that effectively inhibits cellular attachment
  • Surface minimizes protein absorption, enzyme activation and cellular activation
  • Surface is noncytotoxic, biologically inert and nondegradable
  • Sterilized by gamma irradiation
  • Black wall microplates have low background fluorescence, minimal light scatter and reduced crosstalk
  • Opaque walls to prevent well-to-well crosstalk
  • Can be used for both top and bottom reading instruments
  • Bulk packed 10 per bag
  • Novel well geometry aids formation of spheroids in center of well
  • Optically clear round bottom with black opaque microplate body
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
Barcode Includes Barcode
Lid With Lid
Color Black, Clear Bottom
Volume (Metric) Well 90 μL
No. of Wells 384
Volume (Metric) Working 20 to 80 μL
Sterility Sterile
Surface Treatment Ultra-low Binding
Well Shape Round
Quantity 10/Pk., 50/Cs.
WARNING: Cancer - www.P65Warnings.ca.gov