Learn More
Analytical Sales & Services 384-well Cell Culture/Imaging Microplate, 140µL Per Well
Catalog No. 5000806062
Encompass
384-well Cell Culture/Imaging Microplate\n\n• Black Polystyrene\n• Clear, Flat Bottom\n• 140µL Per Well\n• Comes with Lid and Tissue Culture Treatment\n• Irradiated\n\nQty/Pkg: 32
Catalog No. 50-008-06062 Supplier Analytical Sales & Services Supplier No. 384628
384-well Cell Culture/Imaging Microplate\n\n• Black Polystyrene\n• Clear, Flat Bottom\n• 140µL Per Well\n• Comes with Lid and Tissue Culture Treatment\n• Irradiated\n\nQty/Pkg: 32