Learn More
Thomas Scientific 384 Well Cell Culture Plate, Clear, Flat, TC, Sterile
SDP
Catalog No. 501458087
Encompass
Nest cell culture plates have round edges for better handling. Low profile, low-evaporation lid design assure uniform growth. Small feet on the bottom corners of each plate eliminate scraping of the microscope lens. Sterilized by E-beam High clarity, 100% virgin polystyrene Vacuum Plasma tissue culture treatment Clear lot number for batch traceability Peel-to-open, individually wrapped & sterilized by gamma irradiation Markings of well coordinates available for 12, 24, 48 and 96 well plates Non-Pyrogenic, DNase/RNase FREE
This item is not returnable. View return policy
Catalog No. 50-145-8087 Supplier Thomas Scientific Supplier No. 1194Y88
Nest cell culture plates have round edges for better handling. Low profile, low-evaporation lid design assure uniform growth. Small feet on the bottom corners of each plate eliminate scraping of the microscope lens. Sterilized by E-beam High clarity, 100% virgin polystyrene Vacuum Plasma tissue culture treatment Clear lot number for batch traceability Peel-to-open, individually wrapped & sterilized by gamma irradiation Markings of well coordinates available for 12, 24, 48 and 96 well plates Non-Pyrogenic, DNase/RNase FREE