Learn More
Alkali Scientific 384 Well Cell Culture Plate with Lid, Flat, Non-Treated, Sterile, 50/cs
SDP
Catalog No. 502388838
Encompass_Preferred
Cellpro™ Cell Culture Plate come in 384-well and they are constructed of 100% virgin polystyrene and feature a consistent design. The main differences are in terms of the size of the cell growth area and the volume of the wells. They have round edges for better handling and a low profile, low-evaporation lid design to assure uniform growth. They are non-treated. Small feet on the bottom corners of each plate eliminate scraping of the microscope lens. They are non-pyrogenic, DNase/RNase free, and Gamma sterilized. 50 per case.
Catalog No. 50-238-8838 Supplier Alkali Scientific Supplier No. TL5384
Cellpro™ Cell Culture Plate come in 384-well and they are constructed of 100% virgin polystyrene and feature a consistent design. The main differences are in terms of the size of the cell growth area and the volume of the wells. They have round edges for better handling and a low profile, low-evaporation lid design to assure uniform growth. They are non-treated. Small feet on the bottom corners of each plate eliminate scraping of the microscope lens. They are non-pyrogenic, DNase/RNase free, and Gamma sterilized. 50 per case.