Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Falcon™ 384-well, Cell Culture-treated Flat Bottom Microplate PROMO

Catalog No. 08772114S24 Shop All Corning Products
Change view
Click to view available options
Color:
Clear
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
08-772-114S24 Clear
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 08-772-114S24 Supplier Falcon™ Supplier No. 08772114S24

This 384-well, Cell Culture-treated Flat Bottom Microplate is ideal for use in high throughput systems.

  • 384-well microplates are compatible with automation systems and meet standard ANSI/SBS footprint dimensions for microplates
TRUSTED_SUSTAINABILITY

Specifications

Format Standard
Lid With Lid
Content And Storage Ambient Temperature
Culture Area 10 mm2
Array 16 x 24
Binding Type TC-Treated
Color Clear
For Use With (Equipment) Automation Systems
Material Polystyrene
No. of Wells 384
Volume (Metric) Working 15 to 110 μL
Sterility Sterile
Surface Treatment Cell Culture
Well Shape Flat Bottom (F-well)
No. per Case 50 Plates/Cs.
No. per Pack 5 Plates/Pk.
Well Color Clear
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.