Learn More
Analytical Sales & Services 384-Well Collection Plate, 15-145µL Working Volume
Catalog No. 5000804788
Encompass
384-Well Polypropylene Collection Plate, Flat Bottom, 15-145µL Working Volume\n\nQty/Pkg: 100
Catalog No. 50-008-04788 Supplier Analytical Sales & Services Supplier No. 384201
384-Well Polypropylene Collection Plate, Flat Bottom, 15-145µL Working Volume\n\nQty/Pkg: 100