Learn More
Analytical Sales & Services 384-Well Collection Plate, 65µL, Round Openings, Black
Catalog No. 5000804698
Encompass
384-Well Collection Plate\n• Round Wells with Flat Bottoms\n• 65µL/Well\n• Black Polypropylene\n\nQty/Pkg: 100
Catalog No. 50-008-04698 Supplier Analytical Sales & Services Supplier No. 38465B
384-Well Collection Plate\n• Round Wells with Flat Bottoms\n• 65µL/Well\n• Black Polypropylene\n\nQty/Pkg: 100