Learn More
Analytical Sales & Services 384-Well Collection Plates (COC) – Square Well
Catalog No. 5000807022
Encompass
384 Well Plate (COC), Optically Clear to 230nm, 131µL Volume, Flat Bottom\n\nQty/Pkg: 40
Catalog No. 50-008-07022 Supplier Analytical Sales & Services Supplier No. 384801
384 Well Plate (COC), Optically Clear to 230nm, 131µL Volume, Flat Bottom\n\nQty/Pkg: 40