Learn More
CELLTREAT Scientific Products 384 Well Tissue Culture Plate with Lid, Individual, Sterile
SDP
Catalog No. 501656689
Encompass_Preferred
Smaller size wells use less volume, reducing costs. Increase the number of drug screening targets within the standard footprint. Ideal for automated applications, colorimetric assays, compound storage or small volume applications. Cell culture treatment process provides consistent and even hydrophilic surface for improved cell attachment. Polystyrene is optically clear for easy cell viewing and imaging. Plates are labeled with alpha-numeric markings for easy identification on individual wells. Each well has a raised rim which nests into the rings on the single direction lid to reduce evaporation and limit the risk of cross contamination. Lids have a stacking feature for added stability when handling tall stacks of plates. Each plate is printed with lot number for easy identification and tracking. Sturdy individual blister pack prevents damage when handling or shipping. Treated cell growth area: 0.1135cm¿/well. RNase/DNase free and non-pyrogenic. Individually packed sterile, 50/case.
Catalog No. 50-165-6689 Supplier CELLTREAT Scientific Products Supplier No. 229138
Smaller size wells use less volume, reducing costs. Increase the number of drug screening targets within the standard footprint. Ideal for automated applications, colorimetric assays, compound storage or small volume applications. Cell culture treatment process provides consistent and even hydrophilic surface for improved cell attachment. Polystyrene is optically clear for easy cell viewing and imaging. Plates are labeled with alpha-numeric markings for easy identification on individual wells. Each well has a raised rim which nests into the rings on the single direction lid to reduce evaporation and limit the risk of cross contamination. Lids have a stacking feature for added stability when handling tall stacks of plates. Each plate is printed with lot number for easy identification and tracking. Sturdy individual blister pack prevents damage when handling or shipping. Treated cell growth area: 0.1135cm¿/well. RNase/DNase free and non-pyrogenic. Individually packed sterile, 50/case.