Learn More
Greiner Bio-One 384-well F-Bottom Polystyrene Microplates
Catalog No. 07000052 Shop All Greiner Bio-One Products
Change view
Click to view available options
Color:
Black
White
Sterility:
Non-sterile
Sterile
Surface Treatment:
FLUOTRAC 200
FLUOTRAC 600
LUMITRAC 200
LUMITRAC 600
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Sterility Surface Treatment
07-000-052 Black Sterile FLUOTRAC 600
07-000-054 White Sterile LUMITRAC 600
07-000-056 Black Non-sterile FLUOTRAC 200
07-000-058 White Non-sterile LUMITRAC 200
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 07-000-052 Supplier Greiner Bio-One Supplier No. 781077
Add to Cart
Add to Cart
Suitable for luminescence measurements. Greiner Bio-One 384-Well F-Bottom Polystyrene Microplates is free of detectable DNase, RNase, human DNA.
  • Non-Pyrogenic
  • Material: Polystyrene
  • Free of detectable DNase, RNase, human DNA
  • Traced way back to production through defined LOT number system
  • Well-to-well spacing: 4.5mm
Certifications
DIN ISO 9001
TRUSTED_SUSTAINABILITY
Specifications
Format Standard
Lid Without Lid
Array 16 x 24
Color Black
Material Polystyrene
No. of Wells 384
Sterility Sterile
Surface Treatment FLUOTRAC 600
Well Shape Flat Bottom (F-well)
Quantity 40/Cs.