Learn More
Corning™ 384-Well, Cell Culture-Treated, Flat-Bottom, Low Flange Microplate
GSA_VAGREENER_CHOICE
Catalog No. 07201013 Shop All Corning Products
Change view
Click to view available options
Color:
Black, Clear Bottom
Catalog No. Color
07-201-013 Black, Clear Bottom
1 options
This item is not returnable. View return policy
Catalog No. 07-201-013 Supplier Corning™ Supplier No. 3764
  • PRODUCT PROMOTION
    $2,140.00
    $1,205.00 / Case of 100
    Save $935.00
    44% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart
Add to Cart
This item is not returnable. View return policy
Color:Black, Clear Bottom; Sterility:Sterile; Lid:With Lid
  • Opaque walls to prevent well-to-well crosstalk
  • Optically clear flat well bottom permits direct microscopic viewing
  • Can be used for both top and bottom reading instruments
  • Flat bottoms with 112μL total volume
  • Recommended working volumes of 20 to 80μL
  • TC treated for optimal cell attachment
  • Sterilized by gamma radiation and certified nonpyrogenic
  • Individual alphanumeric codes for well identification
  • Bulk packed 20 per bag
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
Barcode No Barcode
Lid With Lid
Array 16 x 24
Color Black, Clear Bottom
Product Type Microplate
Material Polystyrene
No. of Wells 384
Volume (Metric) Working 20 to 80 μL
Sterility Sterile
Surface Treatment Cell Culture
Well Shape Square, Flat Bottom (F-well)
No. per Pack 20/Pk.
Quantity 100/Cs.
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov