Learn More
Corning™ 384 Well Flat Clear Bottom Polystyrene Untreated Microplates, low flange, no lid, nonsterile
GSA_VAGREENER_CHOICE
Catalog No. 07201012 Shop All Corning Products
Change view
Click to view available options
Color:
Black
Catalog No. Color
07-201-012 Black
1 options
This item is not returnable. View return policy
Catalog No. 07-201-012 Supplier Corning™ Supplier No. 3762
Get Up to $2,500 in Products with Purchase
View all promotions
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy
Untreated (medium binding) polystyrene surface is hydrophobic in nature and binds biomolecules through passive interactions. Corning™ 384 Well Flat Clear Bottom Polystyrene Untreated Microplates are ideally suited for fluorescent assays using either top or bottom detection plate readers.
  • Recommended working volume of up to 20 to 80μL
  • Conforms to standard microplate footprint and dimensions
  • Not treated (or medium binding) polystyrene surface is hydrophobic in nature and binds biomolecules through passive interactions
  • Suitable primarily for the immobilization of large molecules, such as antibodies, that have large hydrophobic regions that can interact with the surface
  • Untreated microplates have a binding capacity of approximately 100 to 200ng lgG/cm²
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
Array 16 x 24
Color Black
Material Polystyrene
No. of Wells 384
Volume (Metric) Working 20 to 80 μL
Sterility Non-sterile
Surface Treatment Untreated
Well Shape Square, Flat Bottom (F-well)
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov