Learn More
MatTek Corporation MatTek 384-Well Glass Bottom Plate, No. 1.5 Coverslip, Gamma-Sterilized, 98 mm x 67 mm
Catalog No. NC2052808
Encompass
The MatTek Corporation 384-well format glass bottom plate combines optical clarity with compatibility for high-resolution microscopy applications. With a large viewing area of 98 mm x 67 mm, it is designed for live cell and fluorescence imaging, providing exceptional performance by minimizing backscatter and eliminating well-to-well crosstalk. This product is gamma-sterilized, enhancing its suitability for long-term tissue culture.
Catalog No. NC2052808 Supplier MatTek Corporation Supplier No. P384G1.510872C
The MatTek Corporation 384-well format glass bottom plate combines optical clarity with compatibility for high-resolution microscopy applications. With a large viewing area of 98 mm x 67 mm, it is designed for live cell and fluorescence imaging, providing exceptional performance by minimizing backscatter and eliminating well-to-well crosstalk. This product is gamma-sterilized, enhancing its suitability for long-term tissue culture.