Learn More
Molecular Devices Molecular Devices Microplate Washer, 96/384-Well, Four Inlets, Interchangeable Heads
Catalog No. 5000014707
Encompass_Preferred
The AquaMax 4000 is a fully self-contained automated microplate washer for biochemical and cell-based assays, supporting simultaneous aspiration and dispensing across 96- and 384-well plates and designed for benchtop or robotic integration.
  • Interchangeable 96 and 384 well wash heads for assay flexibility.
  • Four fluid inlets to accommodate multiple reagent lines.
  • Cell wash head options with angled pins for gentle washing of weakly adherent cells.
  • Touchscreen programmable interface for customizable wash protocols.
  • Fully self-contained operation without external pumps or computer.
  • Compatible with SBS-format plates and robotic plate handlers for automation.
Catalog No. 50-000-14707 Supplier Molecular Devices Supplier No. 03105227
Add to Cart
Add to Cart
Request a Quote
The AquaMax 4000 is a fully self-contained automated microplate washer for biochemical and cell-based assays, supporting simultaneous aspiration and dispensing across 96- and 384-well plates and designed for benchtop or robotic integration.
  • Interchangeable 96 and 384 well wash heads for assay flexibility.
  • Four fluid inlets to accommodate multiple reagent lines.
  • Cell wash head options with angled pins for gentle washing of weakly adherent cells.
  • Touchscreen programmable interface for customizable wash protocols.
  • Fully self-contained operation without external pumps or computer.
  • Compatible with SBS-format plates and robotic plate handlers for automation.