Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corning™ 384-Well, Ultra-Low Binding, Flat-Bottom Microplate
GREENER_CHOICE

Catalog No. 07201446 Shop All Corning Products
Change view
Click to view available options
Color:
Black, Clear Bottom
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
07-201-446 Black, Clear Bottom
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07-201-446 Supplier Corning™ Supplier No. 4588
Get Up to $2,500 in Products with Purchase
View all promotions
Add to Cart
Add to Cart
Request a Quote

Color:Black, Clear Bottom; Sterility:Sterile; Lid:With Lid

Suited for fluorescent and luminescent assays using either top or bottom detection microplate readers.

TRUSTED_SUSTAINABILITY

Specifications

Lid With Lid
Culture Area 0.06 cm2
Array 16 x 24
Binding Type Ultra-Low Attachment
Color Black, Clear Bottom
Material Polystyrene
No. of Wells 384
Sterility Sterile
Surface Treatment Ultra-low Binding
Well Shape Flat Bottom (F-well)
No. per Case 100/Cs.
No. per Pack 20/Pk.
Dimensions (L x W x H) 127.8 x 85.5 x 14.35 mm (5 x 3.36 x 0.56 in.)
Depth (Metric) 11.6 mm
Diameter (Metric) Well Top/bottom: 3.63/2.82 mm
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.