Learn More
Description
Suited for fluorescent and luminescent assays using either top or bottom detection microplate readers.
Specifications
Specifications
| Barcode | No Barcode |
| Lid | With Lid |
| Culture Area | 0.06 cm2 |
| Array | 16 x 24 |
| Color | Black, Clear Bottom |
| Product Type | Collagen-coated Microplate |
| Material | Polystyrene |
| No. of Wells | 384 |
| Sterility | Sterile |
| Surface Treatment | Collagen |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.