Learn More
Corning™ 384-Well, Multi-Treated, Flat-Bottom Microplate
Catalog No. 07201447 Shop All Corning Products
Change view
Click to view available options
Color:
Black, Clear Bottom
Catalog No. Color
07-201-447 Black, Clear Bottom
1 options
Catalog No. 07-201-447 Supplier Corning™ Supplier No. 4589
Color:Black, Clear Bottom; Sterility:Sterile; Lid:With Lid
(This is the suggested replacement for 3828)
10 per Bag, with Lid
Black wall plates have low background fluorescence, minimal light scatter and reduced crosstalk Opaque walls to prevent well-to-well crosstalk Optically clear flat well bottom permits direct microscopic viewing Can be used for both top and bottom reading instruments Flat bottoms with 112 μL total volume Recommended working volumes of 20 to 80 μL Individual alphanumeric codes for well identification Six different surface treatments on a single plate (Collagen I, Fibronectin, Gelatin, Laminin, Poly-D-Lysine, TC-treated)
TRUSTED_SUSTAINABILITY
Specifications
Lid With Lid
Array 16 x 24
Color Black, Clear Bottom
Material Polystyrene
No. of Wells 384
Sterility Sterile
Surface Treatment Multi-treated
Well Shape Flat Bottom (F-well)
No. per Case 10/Cs.
No. per Pack 1/Pk.
Dimensions (L x W x H) 127.8 x 85.5 x 14.35 mm (5 x 3.36 x 0.56 in.)
Depth (Metric) 11.6 mm
Diameter (Metric) Well Top/bottom: 3.63/2.82 mm