Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corning™ 384-Well, Non-Treated, Flat-Bottom Microplate
GREENER_CHOICE

Catalog No. 07201441 Shop All Corning Products
Change view
Click to view available options
Color:
White, Clear Bottom
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
07-201-441 White, Clear Bottom
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07-201-441 Supplier Corning™ Supplier No. 3763
Get Up to $2,500 in Products with Purchase
View all promotions
Add to Cart
Add to Cart

Color:White; Sterility:Non-sterile; Lid:Without Lid

Suited for fluorescent and luminescent assays using either top or bottom detection microplate readers.

TRUSTED_SUSTAINABILITY

Specifications

Lid Without Lid
Array 16 x 24
Packaging 25/Pk., 100/Cs.
Color White, Clear Bottom
Material Polystyrene
No. of Wells 384
Sterility Non-sterile
Surface Treatment Non-treated
Well Shape Flat Bottom (F-well)
No. per Case 100/Cs.
No. per Pack 25/Pk.
Dimensions (L x W x H) 127.8 x 85.5 x 14.35 mm (5 x 3.36 x 0.56 in.)
Depth (Metric) 11.6 mm
Diameter (Metric) Well Top/bottom: 3.63/2.82 mm
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.