Learn More
Corning™ 384-Well, Poly-D Lysine-Treated, Flat-Bottom Microplate
Catalog No. 07201444 Shop All Corning Products
Change view
Click to view available options
Color:
Black, Clear Bottom
Catalog No. Color
07-201-444 Black, Clear Bottom
1 options
Catalog No. 07-201-444 Supplier Corning™ Supplier No. 3768
Get Up to $2,500 in Products with Purchase
View all promotions
Add to Cart
Add to Cart
Request a Quote
Color:Black, Clear Bottom; Sterility:Sterile; Lid:With Lid
Suited for fluorescent and luminescent assays using either top or bottom detection microplate readers.
TRUSTED_SUSTAINABILITY
Specifications
Barcode No Barcode
Lid With Lid
Culture Area 0.06 cm2
Array 16 x 24
Color Black, Clear Bottom
Product Type Microplate
Material Polystyrene
No. of Wells 384
Sterility Sterile
Surface Treatment Poly-D-Lysine
Well Shape Flat Bottom (F-well)
No. per Case 100/Cs.
No. per Pack 20/Pk.
Dimensions (L x W x H) 127.8 x 85.5 x 14.35 mm (5 x 3.36 x 0.56 in.)
Depth (Metric) 11.6 mm
Diameter (Metric) Well Top/bottom: 3.63/2.82 mm