Learn More
CELLTREAT Scientific Products 384 Well Non-Treated Plate with Lid, Individual, Sterile
SDP
Supplier:  CELLTREAT Scientific Products 229538
Encompass_Preferred
Smaller size wells use less volume, reducing costs. Increase the number of drug screening targets within the standard footprint. Ideal for automated applications, colorimetric assays, compound storage or small volume applications. Available in tissue culture treated or non-treated versions. Polystyrene is optically clear for easy cell viewing and imaging. Plates are labeled with alpha-numeric markings for easy identification on individual wells. Each well has a raised rim which nests into the rings on the single direction lid to reduce evaporation & limit the risk of cross contamination. Lids have a stacking feature for added stability when handling tall stacks of plates. Each plate is printed with lot number for easy identification and tracking. Sturdy individual blister pack prevents damage when handling or shipping. Working volume: 100 - 110¿L/well. RNase/DNase free & non-pyrogenic. Case is printed with the lot number for quality traceability. Individually packed sterile, 50/case.
Catalog No. 50-165-6692