Learn More
Bellco Glass, Inc. 384 Well Non-Treated Plate
SDP
Catalog No. 502330594
Encompass_Preferred
DescriptionPlates are labeled with alpha-numeric markings for easy identification of individual wellsLids have a stacking feature for added stability when handling tall stacks of plates384 Well Plates are ideal for automated applications, colorimetric assays, compound storage or small volume applications Pack DescriptionIndividualCase Quantity50SterileYesNon-pyrogenicYesPrimary ColorClearPrimary MaterialPolystyreneWorking Volume0.1-0.11mLBottom ShapeFlatTissue Culture TreatedNoRNase/DNase FreeYes
Catalog No. 50-233-0594 Supplier Bellco Glass, Inc. Supplier No. 229538000
DescriptionPlates are labeled with alpha-numeric markings for easy identification of individual wellsLids have a stacking feature for added stability when handling tall stacks of plates384 Well Plates are ideal for automated applications, colorimetric assays, compound storage or small volume applications Pack DescriptionIndividualCase Quantity50SterileYesNon-pyrogenicYesPrimary ColorClearPrimary MaterialPolystyreneWorking Volume0.1-0.11mLBottom ShapeFlatTissue Culture TreatedNoRNase/DNase FreeYes