Learn More
Electron Microscopy Sciences NUNC™ 384 Well Bottom Plate White, Cell Culture, Sterile
SDP
Catalog No. 5033072
Encompass_Preferred
The NUNC Coverglass Optical Bottom 384 Well Plates have an optically clear polystyrene plate bottom. This clear base permits microscopic viewing and reading of well. Can be read from top or bottom. Rounded square wells elimiate wicking. Optimized for scintillation counting. Working range of 10-100 uL/well. White upper structure with Polymer bottom.
Color: White
Type: Cell Culture
Sterile: Sterile
Catalog No. 50-330-72 Supplier Electron Microscopy Sciences Supplier No. 6481025
The NUNC Coverglass Optical Bottom 384 Well Plates have an optically clear polystyrene plate bottom. This clear base permits microscopic viewing and reading of well. Can be read from top or bottom. Rounded square wells elimiate wicking. Optimized for scintillation counting. Working range of 10-100 uL/well. White upper structure with Polymer bottom.
Color: White
Type: Cell Culture
Sterile: Sterile