Learn More
Electron Microscopy Sciences NUNC™ 384 Well Optical Bottom Plate Black, Non-treated
SDP
Catalog No. 5033079
Encompass_Preferred
The NUNC Coverglass Optical Bottom 384 Well Plates have an optically clear polystyrene plate bottom. This clear base permits microscopic viewing and reading of well. Can be read from top or bottom. Rounded square wells elimiate wicking. Optimized for scintillation counting. Working range of 10-100 uL/well. Black upper structure with Polymer bottom.
Color: Black
Type: Non-treated
Catalog No. 50-330-79 Supplier Electron Microscopy Sciences Supplier No. 6481120
The NUNC Coverglass Optical Bottom 384 Well Plates have an optically clear polystyrene plate bottom. This clear base permits microscopic viewing and reading of well. Can be read from top or bottom. Rounded square wells elimiate wicking. Optimized for scintillation counting. Working range of 10-100 uL/well. Black upper structure with Polymer bottom.
Color: Black
Type: Non-treated