Learn More
Electron Microscopy Sciences NUNC™ 384 Well Optical Bottom Plate Black, Poly-D-Lysine
SDP
Supplier:  Electron Microscopy Sciences 6481054
Encompass_Preferred
The NUNC Coverglass Optical Bottom 384 Well Plates have an optically clear polystyrene plate bottom. This clear base permits microscopic viewing and reading of well. Can be read from top or bottom. Rounded square wells elimiate wicking. Optimized for scintillation counting. Working range of 10-100 uL/well. Black upper structure with Polymer bottom. Poly-D-Lysine surfaces for cell-based array. With lid.
Color: Black
Type: Poly-D-Lysine
Sterile: Non-sterile
Catalog No. 50-330-74