Learn More
Electron Microscopy Sciences NUNC™ 384 Well Optical Bottom Plate, Black, Collagen I
SDP
Catalog No. 5033071
Encompass_Preferred
The NUNC Coverglass Optical Bottom 384 Well Plates have an optically clear polystyrene plate bottom. This clear base permits microscopic viewing and reading of well. Can be read from top or bottom. Rounded square wells elimiate wicking. Optimized for scintillation counting. Working range of 10-100 uL/well. black upper structure with Polymer bottom. Collagen I surfaces for cell-based array. With lid.
Color: Black
Type: Collagen I
Sterile: Non-sterile
Catalog No. 50-330-71 Supplier Electron Microscopy Sciences Supplier No. 6481052
The NUNC Coverglass Optical Bottom 384 Well Plates have an optically clear polystyrene plate bottom. This clear base permits microscopic viewing and reading of well. Can be read from top or bottom. Rounded square wells elimiate wicking. Optimized for scintillation counting. Working range of 10-100 uL/well. black upper structure with Polymer bottom. Collagen I surfaces for cell-based array. With lid.
Color: Black
Type: Collagen I
Sterile: Non-sterile