Learn More
Research Products International Corp 384 Well Color Orienter, Vertical, 2 per Package
SDP
Catalog No. 50998936
Encompass
Bright color lanes in vertical orientation clearly define columns and rows on assay plates. Help reduce fatigue and errors when pipetting multiple samples into plates and improve efficiency. Reusable plastic cards fit most standard flat and round bottom plates.
Catalog No. 50-998-936 Supplier Research Products International Corp Supplier No. 247807
Bright color lanes in vertical orientation clearly define columns and rows on assay plates. Help reduce fatigue and errors when pipetting multiple samples into plates and improve efficiency. Reusable plastic cards fit most standard flat and round bottom plates.