Learn More
Bio Rad Laboratories Hard-Shell® 384-Well PCR Plates, thin wall, skirted, black/white #HSP3865, 50/pk
Catalog No. NC2938589
Encompass
Pkg of 50, black shell/white well PCR plate for higher fluorescent signals, rigid 2-component design
Catalog No. NC2938589 Supplier Bio Rad Laboratories Supplier No. HSP3865
Pkg of 50, black shell/white well PCR plate for higher fluorescent signals, rigid 2-component design