Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BRANDTECH™ BRAND™ 384-Well PCR Plates

Catalog No. 13882324 Shop All BRANDTECH Scientific Products
Change view
Click to view available options
Product Type:
Full Skirt, Rigid Plate
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
13-882-324 Full Skirt, Rigid Plate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 13-882-324 Supplier BRANDTECH™ BRAND™ Supplier No. 781348

missing translation for 'validMainframeMsg'

Made with polypropylene material, these plates are resistant to a wide variety of chemicals, including phenols, chloroform, and DMSO and is compatible with most robotic samplers and automated liquid handling systems.

  • Non-sterile, 0.3mL BRAND™ 384-well deep well plate has a square bottom and is manufactured from high-quality virgin polypropylene
  • Excellent for high throughput screening (HTS), assays requiring a mother plate, cell and tissue culture, sample storage, and more
  • Plate is resistant to a wide variety of chemicals, including phenols, chloroform, and DMSO and is compatible with most robotic samplers and automated liquid handling systems
  • Plate withstands temperatures as low as -121ºC, making it an excellent choice for cold-room applications and cryo-storage
  • PP plate is autoclavable at 250ºF (121ºC) for 20 minutes
  • Wells are labeled in a standard alpha-numeric pattern to simplify sample identification, and have notched corners to facilitate orientation
TRUSTED_SUSTAINABILITY

Specifications

Format Standard
Array 16 x 24
Autoclavable Autoclavable
Color Clear
Product Type Full Skirt, Rigid Plate
Skirt Style Full
Material Polypropylene
No. of Wells 384
For Use With (Application) qPCR
Quantity 50/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.