Learn More
Analytical Sales & Services 384 Well Peptide Desalting Plate, 140µL
Catalog No. 5000804470
Encompass
384 Well Peptide Desalting Plate, 140µL\n\n• Works for both hydrophilic and hydrophobic peptides; proprietary low-carbon loading resin\n• Removes urea and other salts from peptide samples for downstream applications\n\nQty/Pkg: Each
Catalog No. 50-008-04470 Supplier Analytical Sales & Services Supplier No. 384PDP140
384 Well Peptide Desalting Plate, 140µL\n\n• Works for both hydrophilic and hydrophobic peptides; proprietary low-carbon loading resin\n• Removes urea and other salts from peptide samples for downstream applications\n\nQty/Pkg: Each