Learn More
Electron Microscopy Sciences 384 Well Plate, deep Well
SDP
Catalog No. 5031455
Encompass_Preferred
The 384-well plate is made of natural polypropylene and is non-sterile. Max volume μl/well 252, working volume μl/well 5 - 240.
Catalog No. 50-314-55 Supplier Electron Microscopy Sciences Supplier No. 63498DW
The 384-well plate is made of natural polypropylene and is non-sterile. Max volume μl/well 252, working volume μl/well 5 - 240.