Learn More
Electron Microscopy Sciences 384 Well Plate, Low Volume
SDP
Catalog No. 5031454
Encompass_Preferred
The 384-well plate is made of natural polypropylene and is non-sterile. Max volume μl/well 58, working volume μl/well 2 - 35.
Catalog No. 50-314-54 Supplier Electron Microscopy Sciences Supplier No. 63498LV
The 384-well plate is made of natural polypropylene and is non-sterile. Max volume μl/well 58, working volume μl/well 2 - 35.