Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ WebSeal 384-Well Non-Coated Plastic Microplates, U-Shaped Well

Catalog No. 03252415 Shop All Thermo Scientific Products
Change view
Click to view available options
Total Volume (Metric) Well:
58 μL
120 μL
252 μL
No. per Pack:
100 Microplates
120 Microplates
25 Microplates
60 Microplates
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Total Volume (Metric) Well No. per Pack
03-252-415 120 μL 120 Microplates
03-252-410 58 μL 25 Microplates
03-252-411 58 μL 100 Microplates
03-252-413 252 μL 60 Microplates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 03-252-415 Supplier Thermo Scientific™ Supplier No. 60180P138

missing translation for 'validMainframeMsg'

For any standard application or assay, you can be sure of excellent solvent resistance and low background noise with these high quality polypropylene 384 well microplates.

Whatever your standard or routine application or assay, you can be sure of excellent solvent resistance and low background noise with Thermo Scientific™ WebSeal 384-Well Microplates. Made of high-quality, GC-tested polypropylene, these microplates are ideal for:

  • Pharmaceutical and industrial QA/QC
  • High-throughput screening (HTS)
  • Sample collection and storage
  • Combinatorial chemistry
  • Liquid phase assays

Chemically and Thermally Resistant Polypropylene

  • Broad chemical compatibility with alcohols, acetonitrile and other common HPLC solvents
  • Will tolerate temperatures from −80° C to 121°C
  • GC-tested to ensure low extractables

Broad Instrument Compatibility

  • SBS and ANSI standard footprint design

Well Formats to Suit Every Application

  • Shallow well and deep well formats
  • U- and V- bottom wells ensure optimal sample recovery
TRUSTED_SUSTAINABILITY

Specifications

Format Standard
Array 16 x 24
Product Type Microplate
Material Polypropylene
No. of Wells 384
Volume (Metric) Working 10 to 100 μL
Sterility Non-sterile
Surface Treatment Non-coated
Total Volume (Metric) Well 120 μL
Well Shape Round Bottom (U-well)
No. per Case 120 Packs
No. per Pack 120 Microplates
Height (Metric) 14.4 mm
Show More Show Less

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.