Learn More
Electron Microscopy Sciences 384 Well Plate, Standard Volume
SDP
Catalog No. 5031453
Encompass_Preferred
The 384-well plate is made of natural polypropylene and is non-sterile. Max volume μl/well 120, working volume μl/well 10 - 100.
Catalog No. 50-314-53 Supplier Electron Microscopy Sciences Supplier No. 63498ST
The 384-well plate is made of natural polypropylene and is non-sterile. Max volume μl/well 120, working volume μl/well 10 - 100.