Learn More
Cellvis 384 Well glass bottom plate with high performance #1.5 cover glass, 20/Pack
Catalog No. NC0473621
Encompass
384 well glass bottom plate. Black polystyrene frame with high performance #1.5 cover glass (0.170±0.005mm), with lid, Individually packed. Designed for high resolution imaging such as confocal microscopy. 20 per Pack.
Catalog No. NC0473621 Supplier Cellvis Supplier No. P38415HN
384 well glass bottom plate. Black polystyrene frame with high performance #1.5 cover glass (0.170±0.005mm), with lid, Individually packed. Designed for high resolution imaging such as confocal microscopy. 20 per Pack.