Learn More
Analytical Sales & Services 384 Well Plates, 65µL, Round Opening, White
Catalog No. 5000806899
Encompass
White Polypropylene 384 Well Plates, 65µL, Round Well, Flat Bottom
Catalog No. 50-008-06899 Supplier Analytical Sales & Services Supplier No. 38465W
White Polypropylene 384 Well Plates, 65µL, Round Well, Flat Bottom