Learn More
Greiner Bio-One 384-well Polypropylene PCR Microplates with Skirt
Catalog No. 07000917 Shop All Greiner Bio-One Products
Change view
Click to view available options
Color:
White
Catalog No. Color
07-000-917 White
1 options
Catalog No. 07-000-917 Supplier Greiner Bio-One Supplier No. 785207
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
View Alternate Product
Features alphanumeric printing for rapid sample identification.
  • Free of detectable DNase, RNase, human DNA
  • Suitable for real-time, quantitative PCR
  • Comprehensive thermal cycler compatibility
  • Packaging labeled with product symbols for easier identification and warehousing
TRUSTED_SUSTAINABILITY
Specifications
Barcode Alphanumeric
Array 16 x 24
Color White
Material Polypropylene
No. of Wells 384
Sterility Non-sterile
Total Volume (Metric) Well 45 μL
Temperature Range (Metric) -80°C to +121°C