Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Greiner Bio-One 384-well Polystyrene Flat Bottom Microplates

Catalog No. 07000051 Shop All Greiner Bio-One Products
Change view
Click to view available options
Color:
Clear
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
07-000-051 Clear
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07-000-051 Supplier Greiner Bio-One Supplier No. 781101
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Free of detectable DNase, RNase, human DNA. Greiner Bio-One 384 Well Polystyrene Flat Bottom Microplates are compatible with standard equipment and automated systems.

  • Manufactured under DIN ISO 9001 guidelines
  • Non-Pyrogenic
  • Number of wells are quadrupled
  • Well-to-well spacing is 4.5mm
  • Compatible with standard equipment and automated systems
  • Rounded square well profile

Service Plan

No

TRUSTED_SUSTAINABILITY

Specifications

Binding Property No Binding
Lid Without Lid
Array 16 x 24
Autoclavable Not Autoclavable
Binding Type Non Binding
Color Clear
Material Polystyrene
No. of Wells 384
Volume (Metric) Working 15 to 110 μL
Sterility Non-sterile
Surface Treatment Non-treated
Total Volume (Metric) Well 131 μL
Well Shape Rounded Square
Dnase Free Yes
Max. RCF 4800 x g (swinging bucket rotor)
No. per Case 100
No. per Pack 10
Quantity 100/Cs.
RNase Free Yes
Temperature Range (Metric) -20°C to +60°C
Well Color Clear
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.