Learn More
Greiner Bio-One 384-well Polystyrene Non-binding Flat Bottom Microplate
GREENER_CHOICE
Catalog No. 07000031 Shop All Greiner Bio-One Products
Change view
Click to view available options
Color:
Black
Black, Clear Bottom
Clear
White
White, Clear Bottom
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
07-000-031 White, Clear Bottom
07-000-890 Black
07-000-892 Black, Clear Bottom
07-000-891 White
07-000-060 Clear
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 07-000-031 Supplier Greiner Bio-One Supplier No. 781903
Add to Cart
Add to Cart
Provides an ultra low non-specific biomolecular binding properties. Greiner Bio-One 384 Well Polystyrene Non-binding Flat Bottom Microplate has long-term surface stability without degradation or leaching.
  • Free of detectable DNase, RNase, human DNA
  • Non-pyrogenic
  • Manufactured under DIN ISO 9001 guidelines
  • Higher assay sensitivity with reduced background
Service Plan
No
TRUSTED_SUSTAINABILITY
Specifications
Format Standard
Array 16 x 24
Binding Type Non Binding
Color White, Clear Bottom
Material Polystyrene
No. of Wells 384
Sterility Non-sterile
Well Shape Flat Bottom (F-well)
No. per Pack 10