Learn More
Greiner Bio-One 384-well Polystyrene Non-binding Flat Bottom Microplate
GREENER_CHOICE
Catalog No. 07000891 Shop All Greiner Bio-One Products
Change view
Click to view available options
Color:
Black
Black, Clear Bottom
Clear
White
White, Clear Bottom
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
07-000-891 White
07-000-890 Black
07-000-892 Black, Clear Bottom
07-000-060 Clear
07-000-031 White, Clear Bottom
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 07-000-891 Supplier Greiner Bio-One Supplier No. 781904
Add to Cart
Add to Cart
Provides an ultra low non-specific biomolecular binding properties. Greiner Bio-One 384 Well Polystyrene Non-binding Flat Bottom Microplate has long-term surface stability without degradation or leaching.
  • Free of detectable DNase, RNase, human DNA
  • Non-pyrogenic
  • Manufactured under DIN ISO 9001 guidelines
  • Higher assay sensitivity with reduced background
Service Plan
No
TRUSTED_SUSTAINABILITY
Specifications
Format Standard
Array 16 x 24
Autoclavable Not Autoclavable
Binding Type Non Binding
Color White
Product Type Microplate
Material Polystyrene
No. of Wells 384
Volume (Metric) Working 15 to 110 μL
Sterility Non-sterile
Total Volume (Metric) Well 131 μL
Well Shape Rounded Square
Max. RCF 4800 x g (swinging bucket rotor)
No. per Case 40
No. per Pack 10
RNase Free Yes
Temperature Range (Metric) -20°C to +60°C
Well Color White