Learn More
Greiner Bio-One 384-well Polystyrene Streptavidin-coated Flat Bottom Microplate
GREENER_CHOICE
Catalog No. 07000903 Shop All Greiner Bio-One Products
Change view
Click to view available options
Color:
Black
Black, Clear Bottom
Clear
White
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
07-000-903 White
07-000-902 Clear
07-000-904 Black, Clear Bottom
07-000-905 Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 07-000-903 Supplier Greiner Bio-One Supplier No. 781995
Add to Cart
Add to Cart
Request a Quote
Pre-blocked and ready-to-use.
  • Manufactured under DIN ISO 9001 guidelines
  • Biotin binding capacity: >1.5ng/well or >6pmol/well
  • Solid bottom
  • Storable at room temperature
  • With lid
TRUSTED_SUSTAINABILITY
Specifications
Format Standard
Array 16 x 24
Autoclavable Not Autoclavable
Color White
Material Polystyrene
No. of Wells 384
Sterility Non-sterile
Surface Treatment Streptavidin
Well Shape Rounded Square
Max. RCF 4800 x g (swinging bucket rotor)
No. per Case 40
No. per Pack 5
Temperature Range (Metric) Room Temperature