Learn More
Analytical Sales & Services 384 Well Protein Desalting Plate, 140µL
Catalog No. 5000804773
Encompass
384 Well Protein Desalting Plate, 140µL\n\n• Proprietary resin for processing proteins with molecular weight >7kDa.\n• Removes salts and other small molecules (>95% ) with molecular weight <1kDa from protein samples\n• Recommend refrigeration at 4-8ºC\n\nQty/Pkg: Each
Catalog No. 50-008-04773 Supplier Analytical Sales & Services Supplier No. 384DP140
384 Well Protein Desalting Plate, 140µL\n\n• Proprietary resin for processing proteins with molecular weight >7kDa.\n• Removes salts and other small molecules (>95% ) with molecular weight <1kDa from protein samples\n• Recommend refrigeration at 4-8ºC\n\nQty/Pkg: Each